missing translation for 'onlineSavingsMsg'
Learn More

Fosfructose trisodium, MedChemExpress

Product Code. 16485066
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16485066 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16485066 Supplier Medchem Express Supplier No. HY106950A
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Fosfructose trisodium (Diphosphofructose trisodium;Esafosfan trisodium;FDP trisodium) is a cytoprotective natural sugar phosphate for the potential treatment of cardiovascular ischemia, sickle cell anemia and asthma. It acts by stimulating anaerobic glycolysis which generates adenosine triphosphate under ischemic conditions.

Fosfructose trisodium (Diphosphofructose trisodium

Specifications

Chemical Name or Material Fosfructose trisodium
Physical Form Solid
CAS 38099-82-0
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C6H11Na3O12P2
For Use With (Application) COVID-19-immunoregulation
Synonym Diphosphofructose trisodiumEsafosfan trisodiumFDP trisodium
Solubility Information H2O : 150 mg/mL (369.40 mM; Need ultrasonic)
SMILES O[C@H](C(COP(O)(O[Na])=O)=O)[C@@H]([C@@H](COP(O[Na])(O[Na])=O)O)O
Molecular Weight (g/mol) 406.06
Formula Weight 406.06
Percent Purity 99.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.