missing translation for 'onlineSavingsMsg'
Learn More

Fosfomycin Calcium, TRC

Product Code. 30011906
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011906 1 g 1g
30008731 100 mg 100mg
30004695 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011906 Supplier Toronto Research Chemicals Supplier No. TRCF7275081G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26016-98-8
Molecular Formula C3 H5 O4 P . Ca
Molecular Weight (g/mol) 176.12
Synonym Phosphonic acid, (1,2-epoxypropyl)-, calcium salt (1:1), (1R,2S)-(-)- (8CI), Phosphonic acid, (3-methyloxiranyl)-, calcium salt (1:1), (2R-cis)-, Phosphonic acid, [(2R,3S)-3-methyloxiranyl]-, calcium salt (1:1) (9CI), Calcium fosfomycin, Calcium phosphomycin, Calcium phosphonomycin, Fosfomycin calcium, Fosfomycin calcium salt, Fosfomycin calcium salt (1:1), Fosmicin, Levo-phosphonomycin calcium salt, Phosphonomycin calcium salt
IUPAC Name calcium;[(2R,3S)-3-methyloxiran-2-yl]-dioxido-oxo-λ^{5}-phosphane
SMILES [Ca+2].C[C@@H]1O[C@@H]1P(=O)([O-])[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C3H7O4P.Ca/c1-2-3(7-2)8(4,5)6;/h2-3H,1H3,(H2,4,5,6);/q;+2/p-2/t2-,3+;/m0./s1
Formula Weight 175.9551
Percent Purity >95%
Chemical Name or Material Fosfomycin calcium
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.