Learn More
Thermo Scientific Chemicals Fosetyl-aluminum
Product Code. 17201196 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. 17201196
Brand: Thermo Scientific Chemicals 467280250
Request Bulk or Custom Format
CAS: 39148-24-8 | C6H15AlO9P3, C6H18AlO9P3 | 351.08 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 39148-24-8
Molecular Formula C6H15AlO9P3
Specifications
Quantity 25 g
Solubility Solubility aq. soln.: 120 g/L (20°C). Other solubilities: practically insoluble in acetonitrile, propylene, glycol
Melting Point >300.0°C
Formula Weight 354.11
InChI Key OIPMQULDKWSNGX-UHFFFAOYSA-N
SMILES [Al+3].CCOP(=O)=O.CCOP(=O)=O.CCOP(=O)=O
IUPAC Name aluminium(3+) tris((phosphooxy)ethane)
Molecular Weight (g/mol) 351.08
Chemical Name or Material Fosetyl-Aluminum