missing translation for 'onlineSavingsMsg'
Learn More

Fosaprepitant dimeglumine, MedChemExpress

Product Code. 16488657
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488657 5 mg 5mg
16426058 10 mg 10mg
16422197 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16488657 Supplier Medchem Express Supplier No. HY14407A
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Fosaprepitant dimeglumine (MK-0517) is a prodrug of Aprepitant (HY-10052). Fosaprepitant dimeglumine is a neurokinin-1 receptor antagonist, which is development for the prevention of chemotherapy-induced nausea and vomiting (CINV).

Fosaprepitant dimeglumine (MK-0517) is a prodrug of Aprepitant (HY-10052). Fosaprepitant dimeglumine is a neurokinin-1 receptor antagonist, which is development for the prevention of chemotherapy-induced nausea and vomiting (CINV).

Specifications

Chemical Name or Material Fosaprepitant dimeglumine
CAS 265121-04-8
Color White
Purity Grade Notes Research
Molecular Formula C37H56F7N6O16P
Quantity 5 mg
Synonym MK-0517 L785298
Solubility Information H2O : 50 mg/mL (49.76 mM; Need ultrasonic)
SMILES O=C1N=C(CN2[C@@H](C3=CC=C(F)C=C3)[C@@H](O[C@H](C)C4=CC(C(F)(F)F)=CC(C(F)(F)F)=C4)OCC2)NN1P(O)(O)=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)CNC.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)CNC
Molecular Weight (g/mol) 1004.83
Formula Weight 1004.83
Percent Purity 98.05%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.