Learn More
Poulten & Graf GmbH FORTUNA™ OPTIMA™ Glass and Metal Syringe with Luer Tip
Product Code. 10311032
Change view
Click to view available options
Volume (Metric):
2 mL
5 mL
30 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10311032 5 mL Each
10126260 30 mL Each
10321032 2 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10311032 Supplier Poulten & Graf GmbH Supplier No. 7 20233
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
Poulten & Graf GmbH FORTUNA™ OPTIMA™ Glass and Metal Syringe with Luer Tip is used to measure and dispense fluids.
  • With Luer tip
  • With steel piston
  • Interchangable
  • For technical use only
Specifications
Autoclavable Autoclavable
Material Neutral Glass, Neutral Glass (Barrel), Nickel-plated Brass (Tip), Steel (Plunger)
Color Clear
Description Glass and Metal Syringe with Luer tip
Latex Free Yes
Product Type Glass and Metal Syringe
Disposable Reusable
Graduations Amber
Model Glass and Metal Syringe
Quantity 1
Volume (Metric) 5 mL
Connection Type Luer Tip
Graduated Yes