Learn More
Formoterol-D6 (Major) (Mixture of Diastereomers), TRC
Specifications
Specifications
| Chemical Name or Material | D,L-Formoterol-D6 (Major) |
| CAS | 1020719-45-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C19H18D6N2O4 |
| Quantity | 10 mg |
| Synonym | Formamide, N-[2-hydroxy-5-[1-hydroxy-2-[[1-[(4-methoxyphenyl)methyl-d2]ethyl-1,2,2,2-d4]amino]ethyl]phenyl]- |
| InChI Formula | InChI=1S/C19H24N2O4/c1-13(9-14-3-6-16(25-2)7-4-14)20-11-19(24)15-5-8-18(23)17(10-15)21-12-22/h3-8,10,12-13,19-20,23-24H,9,11H2,1-2H3,(H,21,22)/i1D3,9D2,13D |
| SMILES | [2H]C([2H])([2H])C([2H])(NCC(O)c1ccc(O)c(NC=O)c1)C([2H])([2H])c2ccc(OC)cc2 |
| Molecular Weight (g/mol) | 350.44 |
| Formula Weight | 350.2113 |
| Show More |