missing translation for 'onlineSavingsMsg'
Learn More

Formic acid hydrazide, 98%

Product Code. p-7022227
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11442707

Brand: Thermo Scientific Alfa Aesar A14901.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 624-84-0 | CH4N2O | 60.056 g/mol

Formic acid hydrazide is used as a precursor in the preparation of 1,2,4-triazole derivatives. It is also used in the synthesis of 6-N-formylamino-12,13-dihydro-1,11-dihydroxy-13-(beta-D-glucopyranosyl)-5H-indolo[2,3-a]-pyrrolo[3,4-c]carbazole-5,7(6H)-dione, which is an anticancer agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Formic acid hydrazide is used as a precursor in the preparation of 1,2,4-triazole derivatives. It is also used in the synthesis of 6-N-formylamino-12,13-dihydro-1,11-dihydroxy-13-(beta-D-glucopyranosyl)-5H-indolo[2,3-a]-pyrrolo[3,4-c]carbazole-5,7(6H)-dione, which is an anticancer agent.

Solubility
Soluble in water, ethanol and acetone.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-84-0
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
MDL Number MFCD00007589
InChI Key XZBIXDPGRMLSTC-UHFFFAOYSA-N
Synonym formylhydrazine, formic acid hydrazide, formic hydrazide, hydrazinecarboxaldehyde, formhydrazide, n-formylhydrazine, carbazaldehyde, formylhydrazide, hydrazine, formyl, formal hydrazine
PubChem CID 12229
IUPAC Name formohydrazide
SMILES C(=O)NN

Specifications

Melting Point 55°C to 59°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 635759
Sensitivity Hygroscopic
Solubility Information Soluble in water,ethanol and acetone.
Formula Weight 60.06
Percent Purity 98%
Chemical Name or Material Formic acid hydrazide
compliance-icons
Product Identifier
  • Formic acid hydrazide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Hydrazinecarboxaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.