accessibility menu, dialog, popup

UserName

NH2HNO

CAS RN 624-84-0

IUPAC Name: formohydrazide
Synonyms: N-formylhydrazine
Molecular Weight (g/mol): 60.06
Molecular Formula: CH4N2O
InChi Key: XZBIXDPGRMLSTC-UHFFFAOYSA-N
SMILES: NNC=O
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results