missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde, 37% w/w, Stabilized with Methanol, Pharmpur™, Scharlab

Product Code. 11394124
Change view
Click to view available options
Quantity:
5 L
25 L
Unit Size:
25 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394124 25 L 25 L
11384124 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394124 Supplier Scharlab Supplier No. FO0010025P
  • ONLINE EXCLUSIVE PRICE
    633.15 EUR
    420.65 EUR / 25 L
    Save 212.50 EUR
    33% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used as a disinfectant and in the synthesis of organic products, embalming liquids, analytical chemistry, photography and the pharmaceutical industry.

  • Synonyms: Formalin solution, Formol, Methanal solution, Methyl aldehyde solution
  • Molecular Formula: CH2O
  • Molar Mass: 30.03g/mol
  • CAS: 50-00-0
  • EIN ECS No.: 200-001-8
  • Ignition Temperature: ∼300°C (pure substance)
  • Vapor Pressure: 1.3hPa (formaldehyde)
  • Acidity: Passes test

Chemical Identifiers

CAS 50-00-0
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Melting Point <-15°C
Density 1.09g/cm3
Boiling Point 93°C to 96°C
Flash Point 62°C
Ignition Residue 0.1% max
Quantity 25 L
Residual Solvents Analysed according to guideline CPMP/ICH/283/95
Stabilizer Approx. 10% methanol
Formula Weight 30.03g/mol
Grade Pharmpur™ USP
Concentration or Composition (by Analyte or Components) 34.5–38.0% (acidimetric assay)
Physical Form Solution
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Formaldehyde, solution 37% w/w, extra pure, Pharmpur, Ph Eur, BP, Usp, stabilized with approx. 10%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Specific target organ toxicity after single exposure Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H370-Causes damage to organs.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: formaldehyde, methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.