Learn More
Thermo Scientific™ Forma™ Freezer Replacement Shelves
Product Code. 11786469 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
13.0 cu. ft. Freezers
17.3 cu. ft. Freezers
23.0 cu. ft. Freezers
28 cu. ft. Freezers
Type:
Shelf Kit
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Type unitSize
11786469 13.0 cu. ft. Freezers Shelf Kit Each
11756479 17.3 cu. ft. Freezers Shelf Kit Each
11766479 23.0 cu. ft. Freezers Shelf Kit Each
11786649 28 cu. ft. Freezers Shelf Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11786469 Supplier Thermo Scientific™ Supplier No. 224134
  • ONLINE EXCLUSIVE PRICE
    210.00 EUR
    202.00 EUR / Pack of 1
    Save 8.00 EUR
    3% Off
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket
Use Thermo Scientific™ Forma™ Freezer Replacement Shelves with Forma 8600 and 900 Series Upright Freezers.
  • Stainless steel
  • Kits include mounting hardware
TRUSTED_SUSTAINABILITY
Specifications
Type Shelf Kit
Capacity (English) 13 cu. ft.
Capacity (Metric) 368.1 L
Style Upright
Color Silver
For Use With (Equipment) 13.0 cu. ft. Freezers