Learn More
Sartorius™ Forceps with Silicone Covered Tips
Product Code. 10542923 Shop All Sartorius Products
Change view
Click to view available options
Length (Metric):
105 mm
160 mm
230 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10542923 160 mm Each
11397504 105 mm Each
10767232 230 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10542923 Supplier Sartorius™ Supplier No. YAW32
  • ONLINE EXCLUSIVE PRICE
    41.60 EUR
    37.50 EUR / Pack of 1
    Save 4.10 EUR
    9% Off
In Stock
For use with balance weights
  • Prevents damage to the surface of weights
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 160 mm
No. per Pack 1 Each
Product Type Forceps with Silicone