Learn More
Cole-Parmer™ Foam-Tipped Short Glassware Brush
Product Code. 15798619 Shop All Cole Parmer Products
Change view
Click to view available options
Length (English):
13 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Unit Size
15798619 13 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15798619 Supplier Cole-Parmer™ Supplier No. 8455214
  • ONLINE EXCLUSIVE PRICE
    16.54 EUR
    15.21 EUR / Pack of 1
    Save 1.33 EUR
    8% Off
View all promotions
Estimated Shipment: 25-11-2026
Foam-Tipped Short Glassware Brush - Excellent for all wide mouth bottles, culture, volumetric graduated, filtering and many other kinds of long flasks, jugs and cylinders. Brushes are tough, yet non-abrasive, and dishwasher safe. It works equally well on both plastic and glass bottles. Use with hot water. No detergents needed.
  • Ergonomically designed wide, flat handles - makes for easy rotating of brush
  • Polypropylene and foam bristles, wire and plastic handle
  • Non-abrasive
  • No detergents needed
  • Dishwasher safe
Specifications
Handle Material Wire, Plastic
For Use With (Application) Cleaning glass and plastic bottles, culture flasks, volumetric flasks, filter flasks etc., jugs and cylinders
Length (English) 13 in.
Length (English) Handle 13 in.
Width (English) Bristle 2 in.