missing translation for 'onlineSavingsMsg'
Learn More

FN-1501, MedChemExpress

Product Code. 16497147
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497147 5 mg 5mg
16475316 100 mg 100mg
16464428 10 mg 10mg
16410747 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16497147 Supplier Medchem Express Supplier No. HY111361

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress FN-1501 is a potent inhibitor of FLT3 and CDK, with IC50s of 2.47, 0.85, 1.96, and 0.28 nM for CDK2/cyclin A, CDK4/cyclin D1, CDK6/cyclin D1 and FLT3, respectively. FN-1501 has anticancer activity.

FN-1501 is a potent inhibitor of FLT3 and CDK, with IC50s of 2.47, 0.85, 1.96, and 0.28 nM for CDK2/cyclin A, CDK4/cyclin D1, CDK6/cyclin D1 and FLT3, respectively. FN-1501 has anticancer activity.

Specifications

Chemical Name or Material FN-1501
Physical Form Solid
CAS 1429515-59-2
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C22H25N9O
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 50 mg/mL (115.88 mM)
SMILES O=C(C1=NNC=C1NC2=NC=NC3=C2C=CN3)NC4=CC=C(CN5CCN(C)CC5)C=C4
Molecular Weight (g/mol) 431.49
Formula Weight 431.49
Percent Purity 99.71%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.