Learn More
Applied Biosystems™ Fmoc-L-Histidine (Trt), 10/Pk.
Product Code. 15939172 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
10 each
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15939172 10 each Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15939172 Supplier Applied Biosystems™ Supplier No. 410778
 Restricted Item
Estimated Shipment: 18-11-2026
These are Amino Acid Cartridges for the ABI™ 431-433 Peptide Synthesizer.
  • For Research Use Only
  • Not for use in diagnostics procedures
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Each package contains 10 cartridges.
Format Cartridge
For Use With (Equipment) 433 A Peptide Synthesizer
For Use With (Application) Protein Sequencing & Peptide Synthesis
Product Type Peptide Synthesis Reagents
Quantity 10 each