Learn More
Applied Biosystems™ Fmoc-L-Glutamic Acid (OtBu), 10/Pk.
Shop All Applied Biosystems Products
Click to view available options
:
433A Peptide Synthesizer
Description
- For Research Use Only
- Not for use in diagnostics procedures
Specifications
Specifications
| Content And Storage | Each package contains 10 cartridges. |
| Format | Cartridge |
| For Use With (Equipment) | 433 A Peptide Synthesizer |
| For Use With (Application) | Protein Sequencing & Peptide Synthesis |
| Product Type | Peptide Synthesis Reagents |
| Quantity | 10 Ea. |