missing translation for 'onlineSavingsMsg'
Learn More

Fmoc-d-3-pyridylalanine, TRC

Product Code. 30179032
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179032 100 mg 100mg
30179476 250 mg 250mg
30179198 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179032 Supplier Toronto Research Chemicals Supplier No. TRCF620228100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142994-45-4
Molecular Formula C23H20N2O4
Molecular Weight (g/mol) 388.42
Synonym (alphaR)-alpha-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]-3-pyridinepropanoic Acid;
SMILES OC(=O)[C@@H](Cc1cccnc1)NC(=O)OCC2c3ccccc3c4ccccc24

Specifications

InChI Formula InChI=1S/C23H20N2O4/c26-22(27)21(12-15-6-5-11-24-13-15)25-23(28)29-14-20-18-9-3-1-7-16(18)17-8-2-4-10-19(17)20/h1-11,13,20-21H,12,14H2,(H,25,28)(H,26,27)/t21-/m1/s1
Quantity 100 mg
Formula Weight 388.1423
Shipping Condition Room Temperature
Chemical Name or Material Fmoc-d-3-pyridylalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.