missing translation for 'onlineSavingsMsg'
Learn More

Fmoc-(3S)-3-amino-1-carboxymethylcaprolactame, TRC

Product Code. 30179239
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179239 1 g 1g
30179004 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179239 Supplier Toronto Research Chemicals Supplier No. TRCF6199851G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142855-79-6
Synonym (3S)-Fmoc-3-amino-1-carboxymethyl-caprolactame∣(3S)-3-[[(9H-fluoren-9-ylmethoxy)carbonyl]amino]hexahydro-2-oxo-1H-azepine-1-acetic Acid

Specifications

Format Neat
Molecular Formula C23H24N2O5
MDL Number MFCD00270208
Molecular Weight (g/mol) 408.45
InChI Key ZYSRRJZAOFHCSC-FQEVSTJZSA-N
SMILES OC(=O)CN1CCCC[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C1=O
IUPAC Name 2-[(3S)-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-2-oxoazepan-1-yl]acetic acid
Quantity 1 g
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.