CAS RN 142855-79-6
CAS RN 142855-79-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fmoc-(3S)-3-amino-1-carboxymethylcaprolactame, TRC
CAS: 142855-79-6 Synonym: (3S)-Fmoc-3-amino-1-carboxymethyl-caprolactame∣(3S)-3-[[(9H-fluoren-9-ylmethoxy)carbonyl]amino]hexahydro-2-oxo-1H-azepine-1-acetic Acid