missing translation for 'onlineSavingsMsg'
Learn More

FMK, MedChemExpress

Product Code. 16490608
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
2mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490608 5 mg 5mg
16468158 10 mg 10mg
16460356 2 mg 2mg
16413766 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16490608 Supplier Medchem Express Supplier No. HY52101A5mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress FMK is a an irreversible RSK2 kinase inhibitor, that covalently modifies the C-terminal kinase domain of RSK.

FMK is a an irreversible RSK2 kinase inhibitor, that covalently modifies the C-terminal kinase domain of RSK.

Specifications

Chemical Name or Material FMK
CAS 821794-92-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C18H19FN4O2
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (292.08 mM; Need ultrasonic)
SMILES FCC(C1=C(C2=C(N=CN=C2N1CCCO)N)C3=CC=C(C=C3)C)=O
Molecular Weight (g/mol) 342.37
Formula Weight 342.37
Percent Purity 95.05%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.