missing translation for 'onlineSavingsMsg'
Learn More

Fluvoxamine, TRC

Product Code. 30007994
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007994 50 mg 50mg
30006988 10 mg 10mg
30009289 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007994 Supplier Toronto Research Chemicals Supplier No. TRCF60350150MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54739-18-3
Molecular Weight (g/mol) 318.33
SMILES COCCCC\C(=N/OCCN)\c1ccc(cc1)C(F)(F)F

Specifications

Molecular Formula C15H21F3N2O2
Quantity 50 mg
Percent Purity >95%
Chemical Name or Material Fluvoxamine, Maleate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.