missing translation for 'onlineSavingsMsg'
Learn More

Fluvastatin sodium, MedChemExpress

Product Code. 16466516
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16466516 100 mg 100mg
16412267 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16466516 Supplier Medchem Express Supplier No. HY14664A
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Fluvastatin sodium (XU 62320) is a first fully synthetic, competitive HMG-CoA reductase inhibitor with an IC50 of 8 nM. Fluvastatin sodium protects vascular smooth muscle cells against oxidative stress through the Nrf2-dependent antioxidant pathway.

Fluvastatin sodium (XU 62320) is a first fully synthetic, competitive HMG-CoA reductase inhibitor with an IC50 of 8 nM. Fluvastatin sodium protects vascular smooth muscle cells against oxidative stress through the Nrf2-dependent antioxidant pathway.

Specifications

Chemical Name or Material Fluvastatin sodium
CAS 93957-55-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H25FNNaO4
Quantity 100 mg
Synonym XU 62-320
Solubility Information H2O : 50 mg/mL (115.35 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (115.35 mM; Need ultrasonic)
SMILES O=C(O[Na])C[C@H](O)C[C@H](O)/C=C/C(N1C(C)C)=C(C2=CC=C(F)C=C2)C3=C1C=CC=C3
Molecular Weight (g/mol) 433.45
Formula Weight 433.45
Percent Purity 99.48%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.