missing translation for 'onlineSavingsMsg'
Learn More

Fluticasone 17Beta-Carbonylsulfenic Acid 17-Propionate, TRC

Product Code. 30138388
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138388 100 mg 100mg
17962395 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138388 Supplier Toronto Research Chemicals Supplier No. TRCF599480100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 948566-12-9
Molecular Formula C24 H30 F2 O6 S
Molecular Weight (g/mol) 484.55
Synonym Fluticasone Propionate Imp. B (EP), 6alpha,9-Difluoro-11beta-hydroxy-16alpha-methyl-3-oxo-17-(propanoyloxy)androsta-1,4-dien-17beta-carbo(thioperoxoic) SO-Acid, [[6alpha,9-Difluoro-11beta-hydroxy-16alpha-methyl-3-oxo-17-(propanoyloxy)androsta-1,4-dien-17beta-yl]carbonyl]sulphenic Acid
IUPAC Name [(6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-difluoro-11-hydroxy-17-hydroxysulfanylcarbonyl-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] propanoate
SMILES CCC(=O)O[C@@]1([C@H](C)C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)SO

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C24H30F2O6S/c1-5-19(29)32-24(20(30)33-31)12(2)8-14-15-10-17(25)16-9-13(27)6-7-21(16,3)23(15,26)18(28)11-22(14,24)4/h6-7,9,12,14-15,17-18,28,31H,5,8,10-11H2,1-4H3/t12-,14+,15+,17+,18+,21+,22+,23+,24+/m1/s1
Formula Weight 484.1731
Percent Purity >95%
Chemical Name or Material Fluticasone 17beta-Carbonylsulfenic Acid 17-Propionate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.