Learn More
Flutianil, TRC
Product Code. 30004096
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004096 5 mg 5mg
30009226 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30004096 Supplier Toronto Research Chemicals Supplier No. TRCF5994255MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 958647-10-4
Molecular Formula C19 H14 F4 N2 O S2
Molecular Weight (g/mol) 426.45
Synonym Acetonitrile, 2-[[2-fluoro-5-(trifluoromethyl)phenyl]thio]-2-[3-(2-methoxyphenyl)-2-thiazolidinylidene]-, (2Z)-, (2Z)-2-[[2-Fluoro-5-(trifluoromethyl)phenyl]thio]-2-[3-(2-methoxyphenyl)-2-thiazolidinylidene]acetonitrile, Flutianil, OK 5203, V 10118
IUPAC Name (2Z)-2-[2-fluoro-5-(trifluoromethyl)phenyl]sulfanyl-2-[3-(2-methoxyphenyl)-1,3-thiazolidin-2-ylidene]acetonitrile
SMILES COc1ccccc1N2CCS/C/2=C(\Sc3cc(ccc3F)C(F)(F)F)/C#N
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C19H14F4N2OS2/c1-26-15-5-3-2-4-14(15)25-8-9-27-18(25)17(11-24)28-16-10-12(19(21,22)23)6-7-13(16)20/h2-7,10H,8-9H2,1H3/b18-17-
Formula Weight 426.048
Chemical Name or Material Flutianil