Learn More
Flurprimidol, TRC
Product Code. 30135544
Change view
Click to view available options
Quantity:
25 mg
250 mg
500 mg
Unit Size:
250mg
25mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135544 500 mg 500mg
30135465 25 mg 25mg
17937139 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135544 Supplier Toronto Research Chemicals Supplier No. TRCF598825500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 56425-91-3
Molecular Formula C15 H15 F3 N2 O2
Molecular Weight (g/mol) 312.29
Synonym α-(1-Methylethyl)-α-[4-(trifluoromethoxy)phenyl]-5-pyrimidinemethanol, Cutless, EL 500, Flurprimidol, Topflor, α-Isopropyl-α-[p-(trifluoromethoxy)phenyl]-5-pyrimidinemethanol
IUPAC Name 2-methyl-1-pyrimidin-5-yl-1-[4-(trifluoromethoxy)phenyl]propan-1-ol
SMILES CC(C)C(O)(c1ccc(OC(F)(F)F)cc1)c2cncnc2
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C15H15F3N2O2/c1-10(2)14(21,12-7-19-9-20-8-12)11-3-5-13(6-4-11)22-15(16,17)18/h3-10,21H,1-2H3
Formula Weight 312.1086
Chemical Name or Material Flurprimidol