missing translation for 'onlineSavingsMsg'
Learn More

Flurprimidol, TRC

Product Code. 30135465
Change view
Click to view available options
Quantity:
25 mg
250 mg
500 mg
Unit Size:
250mg
25mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135465 25 mg 25mg
30135544 500 mg 500mg
17937139 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135465 Supplier Toronto Research Chemicals Supplier No. TRCF59882525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56425-91-3
Molecular Formula C15 H15 F3 N2 O2
Molecular Weight (g/mol) 312.29
Synonym α-(1-Methylethyl)-α-[4-(trifluoromethoxy)phenyl]-5-pyrimidinemethanol, Cutless, EL 500, Flurprimidol, Topflor, α-Isopropyl-α-[p-(trifluoromethoxy)phenyl]-5-pyrimidinemethanol
IUPAC Name 2-methyl-1-pyrimidin-5-yl-1-[4-(trifluoromethoxy)phenyl]propan-1-ol
SMILES CC(C)C(O)(c1ccc(OC(F)(F)F)cc1)c2cncnc2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C15H15F3N2O2/c1-10(2)14(21,12-7-19-9-20-8-12)11-3-5-13(6-4-11)22-15(16,17)18/h3-10,21H,1-2H3
Formula Weight 312.1086
Chemical Name or Material Flurprimidol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.