Learn More
Flurofamide, TRC
Product Code. 30240967
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240967 50 mg 50mg
30240753 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30240967 Supplier Toronto Research Chemicals Supplier No. TRCF59668550MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 70788-28-2
Molecular Formula C7H9FN3O2P
Molecular Weight (g/mol) 217.14
Synonym 4-Fluoro-N-(diaminophosphinyl)benzamide, Flurfamide, NSC 342995
SMILES NP(=O)(N)NC(=O)c1ccc(F)cc1
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C7H9FN3O2P/c8-6-3-1-5(2-4-6)7(12)11-14(9,10)13/h1-4H,(H5,9,10,11,12,13)
Formula Weight 217.0416
Chemical Name or Material Flurofamide