missing translation for 'onlineSavingsMsg'
Learn More

Flurbiprofen, 99.6%, MP Biomedicals™

Product Code. 11460596 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460596 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11460596 Supplier MP Biomedicals Supplier No. 0215816325

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 5104-49-4
Color White
Assay Percent Range 99.6%
Molecular Formula C15H13FO2
MDL Number MFCD00079303
Quantity 25 g
Synonym flurbiprofen, ansaid, froben, antadys, cebutid, flurofen, anside, flurbiprofene, flurbiprofeno, flurbiprofenum
InChI Key SYTBZMRGLBWNTM-UHFFFAOYNA-N
SMILES CC(C(O)=O)C1=CC=C(C(F)=C1)C1=CC=CC=C1
IUPAC Name 2-{2-fluoro-[1,1'-biphenyl]-4-yl}propanoic acid
Molecular Weight (g/mol) 244.27
PubChem CID 3394
ChEBI CHEBI:5130
Formula Weight 244.26
Physical Form Powder
Chemical Name or Material Flurbiprofen
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.