Learn More
Flupyradifurone, TRC
Product Code. 30132648
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132648 25 mg 25mg
17992155 1 mg 1mg
17934005 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30132648 Supplier Toronto Research Chemicals Supplier No. TRCF59861525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 951659-40-8
Molecular Formula C12 H11 Cl F2 N2 O2
Molecular Weight (g/mol) 288.68
Synonym 2(5H)-Furanone, 4-[[(6-chloro-3-pyridinyl)methyl](2,2-difluoroethyl)amino]-, 3-[[(6-Chloro-3-pyridyl)methyl](2,2-difluoroethyl)amino]-2H-furan-5-one, 4-[[(6-Chloropyridin-3-yl)methyl](2,2-difluoroethyl)amino]furan-2(5H)-one, BYI 02960, Flupyradifurone
IUPAC Name 3-[(6-chloropyridin-3-yl)methyl-(2,2-difluoroethyl)amino]-2H-furan-5-one
SMILES FC(F)CN(Cc1ccc(Cl)nc1)C2=CC(=O)OC2
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C12H11ClF2N2O2/c13-10-2-1-8(4-16-10)5-17(6-11(14)15)9-3-12(18)19-7-9/h1-4,11H,5-7H2
Formula Weight 288.0477
Percent Purity >95%
Chemical Name or Material Flupyradifurone
Grade HPLC