Learn More
Flupyradifurone 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30315310
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30315310 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30315310 Supplier Dr. Ehrenstorfer Supplier No. DREA13802300AL100
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Flupyradifurone
CAS 951659-40-8
Analyte or Component Names Flupyradifurone
Shipping Condition +20°C
Molecular Formula C12 H11 Cl F2 N2 O2
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 2(5H)-Furanone, 4-[[(6-chloro-3-pyridinyl)methyl](2,2-difluoroethyl)amino]-, 3-[[(6-Chloro-3-pyridyl)methyl](2,2-difluoroethyl)amino]-2H-furan-5-one, 4-[[(6-Chloropyridin-3-yl)methyl](2,2-difluoroethyl)amino]furan-2(5H)-one, BYI 02960, Flupyradifurone
InChI Formula InChI=1S/C12H11ClF2N2O2/c13-10-2-1-8(4-16-10)5-17(6-11(14)15)9-3-12(18)19-7-9/h1-4,11H,5-7H2
SMILES FC(F)CN(Cc1ccc(Cl)nc1)C2=CC(=O)OC2
IUPAC Name 3-[(6-chloropyridin-3-yl)methyl-(2,2-difluoroethyl)amino]-2H-furan-5-one
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 288.68
Quantity 1 mL
Formula Weight 288.0477 g/mol