missing translation for 'onlineSavingsMsg'
Learn More

Fluoxetine, TRC

Product Code. 30006081
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006081 1 g 1g
17982915 100 mg 100mg
17954945 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006081 Supplier Toronto Research Chemicals Supplier No. TRCF5971831G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54910-89-3
Molecular Formula C17 H18 F3 N O
Molecular Weight (g/mol) 309.33
Synonym Benzenepropanamine, N-methyl-γ-[4-(trifluoromethyl)phenoxy]-, Benzenepropanamine, N-methyl-γ-[4-(trifluoromethyl)phenoxy]-, (±)-, N-Methyl-γ-[4-(trifluoromethyl)phenoxy]benzenepropanamine, (±)-Fluoxetine, (±)-N-Methyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propylamine, 3-(p-Trifluoromethylphenoxy)-N-methyl-3-phenylpropylamine, Daforin, Deprex, Fluoxetin Ratiopharm, Fluoxetine, Fluoxin, Fluval, N-Methyl-3-(p-trifluoromethylphenoxy)-3-phenylpropylamine, N-Methyl-3-[4-(trifluoromethyl)phenoxy]-3-phenylpropanamine, NSC 283480, Nikomed, Seronil, Symbiax, dl-3-(p-Trifluoromethylphenoxy)-N-methyl-3-phenylpropylamine
IUPAC Name N-methyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine
SMILES CNCCC(Oc1ccc(cc1)C(F)(F)F)c2ccccc2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C17H18F3NO/c1-21-12-11-16(13-5-3-2-4-6-13)22-15-9-7-14(8-10-15)17(18,19)20/h2-10,16,21H,11-12H2,1H3
Formula Weight 309.134
Percent Purity >95%
Chemical Name or Material Fluoxetine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.