missing translation for 'onlineSavingsMsg'
Learn More

Fluoxetine Hydrochloride, TRC

Product Code. 17952815
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17952815 100 mg 100mg
17963935 50 mg 50mg
17952715 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17952815 Supplier Toronto Research Chemicals Supplier No. TRCF597100100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56296-78-7
Molecular Weight (g/mol) 345.79
Synonym Fluoxetine Hydrochloride, N-Methyl-γ-[4-(trifluoromethyl)phenoxy]benzenepropanamine hydrochloride (1:1), (±)-N-Methyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propylamine hydrochloride, Adofen, Affectine, Alzac 20, Ansilan, Deproxin, Digassim, Erocap, Fluctin, Fluctine, Fludac, Flufran, Flunil, Fluox-Puren, Fluoxac, Fluoxeren, Fluoxetine hydrochloride, Fluoxil, Flutin, Flutine, Fluxen, Fluxetyl, Fluxil, Fontex, Foxetin, LY 110140, Lilly 110140, Lorien, Lovan, Margrilan, Modipran, N-Methyl-3-(4-trifluoromethylphenoxy)-3-phenylpropylamine hydrochloride, Neupax, Nodepe, Nopres, Nuzak, Octozac, Oxedep, Pluzac, Pragmaten, Prizma, Proctin, Prodep, Profluzac, Prozac, Prozac 20, Reconcile, Reneuron, Rowexetina, Salipax, Sanzur, Sarafem, Sinzac, Zactin, Zedprex, Zepax
IUPAC Name N-methyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine;hydrochloride
SMILES Cl.CNCCC(Oc1ccc(cc1)C(F)(F)F)c2ccccc2

Specifications

Molecular Formula C17 H18 F3 N O . Cl H
Quantity 100 mg
InChI Formula InChI=1S/C17H18F3NO.ClH/c1-21-12-11-16(13-5-3-2-4-6-13)22-15-9-7-14(8-10-15)17(18,19)20;/h2-10,16,21H,11-12H2,1H3;1H
Formula Weight 345.1107
Percent Purity >95%
Chemical Name or Material Fluoxetine Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.