missing translation for 'onlineSavingsMsg'
Learn More

Fluoxetine EP Impurity B (as Hydrochloride), Mikromol

Product Code. 30459947
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30459947 100 mg -
30460091 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30459947 Supplier Mikromol Supplier No. MM0256.02

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Fluoxetine EP Impurity B (as Hydrochloride), Mikromol
TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material 1M-3PP HCl (Methyl-3-phenylpropylamine Hydrochloride)
CAS 30684-07-2
Analyte or Component Names 1M-3PP HCl (Methyl-3-phenylpropylamine Hydrochloride)
Shipping Condition Room Temperature
Molecular Formula C10 H15 N . Cl H
Certifications/Compliance ISO 17025
Synonym N-Methyl-3-phenylpropan-1-amine Hydrochloride (Methyl-3-phenylpropylamine Hydrochloride), Fluoxetine Hydrochloride Imp. B (EP), Fluoxetine Imp. B (EP), Fluoxetine USP Related Compound B, Fluoxetine USP RC B, N-Methyl-3-phenylpropan-1-amine Hydrochloride, Fluoxetine Hydrochloride Imp. B (EP) as Hydrochloride, Methyl-3-phenylpropylamine Hydrochloride, 1M-3PP HCl, Fluoxetine Related Compound B as Hydrochloride, Fluoxetine Hydrochloride Impurity B as Hydrochloride, Fluoxetine Impurity B as Hydrochloride
InChI Formula InChI=1S/C10H15N.ClH/c1-11-9-5-8-10-6-3-2-4-7-10;/h2-4,6-7,11H,5,8-9H2,1H3;1H
Solution Type Neat
SMILES Cl.CNCCCc1ccccc1
IUPAC Name N-methyl-3-phenylpropan-1-amine;hydrochloride
Molecular Weight (g/mol) 185.69
Quantity 100 mg
Formula Weight 185.0971 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.