Learn More
Cytiva Sample loops
Product Code. 11340042 Shop All Cytiva Products
Change view
Click to view available options
Volume (Metric):
10 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11340042 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11340042 Supplier Cytiva Supplier No. 18116124
Estimated Shipment: 10-12-2026
Fitted FEP tubing sample loops. Cytiva Sample loops supplied with ferrules and unions.
  • Tubing i.d. 1.6 mm (0.063 inch) o.d. 1/8 in. fitted with nut 5/16 in. male and ferrules
  • 2 x Union UNF 5/16 in. female-1/16 in. male supplied with the sample loop
  • Fluoroethenepropene (FEP)
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 10 mL