missing translation for 'onlineSavingsMsg'
Learn More

Fluoroacetamide, MP Biomedicals

Product Code. 11463185 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463185 10 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11463185 Supplier MP Biomedicals Supplier No. 0521387805

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 640-19-7
Molecular Formula C2H4FNO
MDL Number MFCD00008026
Quantity 10 g
Synonym fluoroacetamide, acetamide, 2-fluoro, monofluoroacetamide, fluorkill, fluoroacetic acid amide, megatox, fussol, yanock, fluoroakil 100, fluorakil 100
InChI Key FVTWJXMFYOXOKK-UHFFFAOYSA-N
SMILES NC(=O)CF
IUPAC Name 2-fluoroacetamide
Molecular Weight (g/mol) 77.06
PubChem CID 12542
ChEBI CHEBI:53124
Formula Weight 77.06
Chemical Name or Material Fluoroacetamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.