missing translation for 'onlineSavingsMsg'
Learn More

Fluorescein Sodium, C.I. 45350, Extra Pure, Scharlab

Product Code. 11334124
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11334124 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11334124 Supplier Scharlab Supplier No. FL01250250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in analytical chemistry as an indicator.

  • Synonyms: 3',6'-Dihydroxyspiro-[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one, Resorcinolphthalein
  • Molecular Formula: C20H10Na2O5
  • Molar Mass: 376.28g/mol
  • CAS: 518-47-8
  • EIN ECS No.: 208-253-0
  • Solubility in water: 500g/l at 20°C
  • Absorption maximum (pH=8.0): 490–492nm
  • Absorptivity (A1%/1cm; l max; 0.0005%, pH 8.0 on dried sample): 1950–2150
  • Acriflavine: Passes test
  • TLC Test: Passes test

Specifications

CAS 518-47-8
Melting Point >360°C
pH 7.0 to 9.0 (2%, H2O)
Molecular Formula C20H10Na2O5
MDL Number MFCD00167039
Quantity 250 g
Synonym 3',6'-Dihydroxyspiro-[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one, Resorcinolphthalein
InChI Key NJDNXYGOVLYJHP-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12
IUPAC Name disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Molecular Weight (g/mol) 376.28
Formula Weight 376.28g/mol
Grade Extra Pure
Concentration or Composition (by Analyte or Components) 90% min (assay, titration with HClO4)
Show More Show Less
compliance-icons
Product Identifier
  • Fluorescein, C.I. 45350, extra pure
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312+H332-Harmful in contact with skin or if inhaled
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P321-Specific treatment (see on this label).
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.