missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluorescein, Laser Grade 99%, Thermo Scientific Chemicals
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 314.0°C to 316.0°C |
| Assay Percent Range | 99% |
| Fluorescence Max | 518nm (in basic CH3OH) |
| Beilstein | 19, 222 |
| Merck Index | 15, 4192 |
| Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in dilute alkalies and dilute acids, soluble in boiling alcohol and ether, insoluble in benzene and chloroform |
| Infrared Spectrum | Authentic |
| Lambda Max | 498nm (in basic MeOH) |
| Formula Weight | 332.3 |
| Percent Purity | 99% |
| Show More |
Safety and Handling
Product Identifier
- Fluorescein
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Fluorescein
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction