missing translation for 'onlineSavingsMsg'
Learn More

Fluorene-1-carboxylic acid, 97%, Thermo Scientific™

Product Code. 11435977
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11435977 25 g 25g
11415977 1 g 1g
11425977 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11435977 Supplier Thermo Scientific Alfa Aesar Supplier No. B25020.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6276-03-5
Molecular Formula C14H10O2
Molecular Weight (g/mol) 210.23
MDL Number MFCD00001130
InChI Key HTPXFGUCAUTOEL-UHFFFAOYSA-N
Synonym 1-fluorenecarboxylic acid, fluorene-1-carboxylic acid, fluorenecarboxylic acid, unii-ao792vd660, 1-fluorene carboxylic acid, 9h-fluorene-1-carboxylicacid, htpxfgucautoel-uhfffaoysa
PubChem CID 80468
IUPAC Name 9H-fluorene-1-carboxylic acid
SMILES C1C2=CC=CC=C2C3=C1C(=CC=C3)C(=O)O

Specifications

Melting Point 246°C to 248°C
Odor Odorless
Quantity 25 g
Formula Weight 210.23
Percent Purity 97%
Chemical Name or Material Fluorene-1-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.