Learn More
Fluoren-3-ol, TRC
Product Code. 30012696
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012696 100 mg 100mg
30011792 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30012696 Supplier Toronto Research Chemicals Supplier No. TRCF462460100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6344-67-8
Molecular Formula C13 H10 O
Molecular Weight (g/mol) 182.22
Synonym 9H-Fluoren-3-ol, Fluoren-3-ol (6CI,7CI,8CI), 3-Hydroxyfluorene, NSC 51318
IUPAC Name 9H-fluoren-3-ol
SMILES Oc1ccc2Cc3ccccc3c2c1
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C13H10O/c14-11-6-5-10-7-9-3-1-2-4-12(9)13(10)8-11/h1-6,8,14H,7H2
Formula Weight 182.0732
Percent Purity >95%
Chemical Name or Material Fluoren-3-ol
Grade HPLC