missing translation for 'onlineSavingsMsg'
Learn More

Flunixin, TRC

Product Code. 18041750
Change view
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
250mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18041750 50 mg 50mg
18082630 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18041750 Supplier Toronto Research Chemicals Supplier No. TRCF45540850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38677-85-9
Molecular Formula C14 H11 F3 N2 O2
Molecular Weight (g/mol) 296.24
Synonym Flunixin, 3-Pyridinecarboxylic acid, 2-[[2-methyl-3-(trifluoromethyl)phenyl]amino]-, 2-[[2-Methyl-3-(trifluoromethyl)phenyl]amino]-3-pyridinecarboxylic acid, 2-(2-Methyl-3-trifluoromethylanilino)nicotinic acid, Flunixin, Sch 14714
IUPAC Name 2-[2-methyl-3-(trifluoromethyl)anilino]pyridine-3-carboxylic acid
SMILES Cc1c(Nc2ncccc2C(=O)O)cccc1C(F)(F)F

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C14H11F3N2O2/c1-8-10(14(15,16)17)5-2-6-11(8)19-12-9(13(20)21)4-3-7-18-12/h2-7H,1H3,(H,18,19)(H,20,21)
Formula Weight 296.0773
Percent Purity >95%
Chemical Name or Material Flunixin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.