Learn More
Flunixin Meglumine, TRC
Product Code. 30134244
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134244 100 mg 100mg
30004883 10 mg 10mg
30011185 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134244 Supplier Toronto Research Chemicals Supplier No. TRCF455400100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 42461-84-7
Molecular Formula C14 H11 F3 N2 O2 . C7 H17 N O5
Molecular Weight (g/mol) 491.46
Synonym Flunixin Meglumine
IUPAC Name (2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol;2-[2-methyl-3-(trifluoromethyl)anilino]pyridine-3-carboxylic acid
SMILES CNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO.Cc1c(Nc2ncccc2C(=O)O)cccc1C(F)(F)F
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C14H11F3N2O2.C7H17NO5/c1-8-10(14(15,16)17)5-2-6-11(8)19-12-9(13(20)21)4-3-7-18-12;1-8-2-4(10)6(12)7(13)5(11)3-9/h2-7H,1H3,(H,18,19)(H,20,21);4-13H,2-3H2,1H3/t;4-,5+,6+,7+/m.0/s1
Formula Weight 491.1879
Percent Purity >95%
Chemical Name or Material Flunixin meglumine
Grade HPLC