Learn More
Fluindapyr, TRC
Product Code. 30133342
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133342 100 mg 100mg
17960455 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30133342 Supplier Toronto Research Chemicals Supplier No. TRCF138380100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1383809-87-7
Molecular Formula C18 H20 F3 N3 O
Molecular Weight (g/mol) 351.3661
Synonym 3-(Difluoromethyl)-N-(7-fluoro-2,3-dihydro-1,1,3-trimethyl-1H-inden-4-yl)-1-methyl-1H-pyrazole-4-carboxamide, 3-(Difluoromethyl)-N-(7-fluoro-1,1,3-trimethyl-2,3-dihydro-1H-inden-4-yl)-1-methyl-1H-pyrazole-4-carboxamide, Fluindapyr
IUPAC Name 3-(difluoromethyl)-N-(7-fluoro-1,1,3-trimethyl-2,3-dihydroinden-4-yl)-1-methylpyrazole-4-carboxamide
SMILES CC1CC(C)(C)c2c(F)ccc(NC(=O)c3cn(C)nc3C(F)F)c12
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C18H20F3N3O/c1-9-7-18(2,3)14-11(19)5-6-12(13(9)14)22-17(25)10-8-24(4)23-15(10)16(20)21/h5-6,8-9,16H,7H2,1-4H3,(H,22,25)
Formula Weight 351.1558
Percent Purity >95%
Chemical Name or Material Fluindapyr
Grade HPLC