Learn More
Flufenacet-ethane sulfonic acid (ESA) sodium 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30326534
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30326534 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30326534 Supplier Dr. Ehrenstorfer Supplier No. DREA13711017AL100
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Flufenacet ethane sulfonic acid ESA sodium salt
CAS 947601-87-8
Analyte or Component Names Flufenacet ethane sulfonic acid ESA sodium salt
Shipping Condition Room Temperature
Molecular Formula C11 H13 F N O4 S . Na
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C11H14FNO4S.Na/c1-8(2)13(11(14)7-18(15,16)17)10-5-3-9(12)4-6-10;/h3-6,8H,7H2,1-2H3,(H,15,16,17);/q;+1/p-1
SMILES [Na+].CC(C)N(C(=O)CS(=O)(=O)[O-])c1ccc(F)cc1
IUPAC Name sodium;2-(4-fluoro-N-propan-2-ylanilino)-2-oxoethanesulfonate
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 297.28
Quantity 1 mL
Formula Weight 297.0447 g/mol