missing translation for 'onlineSavingsMsg'
Learn More

Fluazolate, TRC

Product Code. 17992245
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17992245 5 mg 5mg
17991715 10 mg 10mg
17931205 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17992245 Supplier Toronto Research Chemicals Supplier No. TRCF4075005MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 174514-07-9
Molecular Formula C15 H12 Br Cl F4 N2 O2
Molecular Weight (g/mol) 443.62
Synonym Benzoic acid, 5-[4-bromo-1-methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]-2-chloro-4-fluoro-, 1-methylethyl ester, Fluazolate, Isopropazol, Isopropyl 5-[4-bromo-1-methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]-2-chloro-4-fluorobenzoate, JV 485, MON 48500, Twin-Agro
IUPAC Name propan-2-yl 5-[4-bromo-1-methyl-5-(trifluoromethyl)pyrazol-3-yl]-2-chloro-4-fluorobenzoate
SMILES CC(C)OC(=O)c1cc(c(F)cc1Cl)c2nn(C)c(c2Br)C(F)(F)F

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C15H12BrClF4N2O2/c1-6(2)25-14(24)7-4-8(10(18)5-9(7)17)12-11(16)13(15(19,20)21)23(3)22-12/h4-6H,1-3H3
Formula Weight 441.9707
Percent Purity >95%
Chemical Name or Material Fluazolate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.