missing translation for 'onlineSavingsMsg'
Learn More

Fluazolate-d3, TRC

Product Code. 30405499
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30405499 10 mg
30399958 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30405499 Supplier Toronto Research Chemicals Supplier No. TRCF40750210MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1189932-72-6
Molecular Formula C15H9D3BrClF4N2O2
Molecular Weight (g/mol) 446.64
SMILES [2H]C([2H])([2H])n1nc(c(Br)c1C(F)(F)F)c2cc(C(=O)OC(C)C)c(Cl)cc2F

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C15H12BrClF4N2O2/c1-6(2)25-14(24)7-4-8(10(18)5-9(7)17)12-11(16)13(15(19,20)21)23(3)22-12/h4-6H,1-3H3/i3D3
Chemical Name or Material Fluazolate-d3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.