Learn More
Brady™ Floor Stand
Product Code. 11796655 Shop All Brady Products
Change view
Click to view available options
Displayed Text:
ATTENTION SOL GLISSANT
ATTENTION ZONE DE TRAVAIL
ENTREE INTERDITE
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Displayed Text Unit Size
11796655 ATTENTION SOL GLISSANT Each
11712005 ENTREE INTERDITE Each
11706665 ATTENTION ZONE DE TRAVAIL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11796655 Supplier Brady™ Supplier No. 256262
  • ONLINE EXCLUSIVE PRICE
    80.35 EUR
    74.94 EUR / Pack of 1
    Save 5.41 EUR
    6% Off
In Stock
Floor Stand designed to warn for hazardous areas, putting valuable safety information directly at the point of need.
  • These floor signs are manufactured from polyester, with a clear matt anti-slip laminate to prevent falling accidents while protecting the graphics underneath.
Specifications
Displayed Text ATTENTION SOL GLISSANT
Height (English) 25.98 in.
Height (Metric) 660 mm
Shape Rectangle
Width (English) 17.72 in.
Width (Metric) 450 mm
Color Black, Red on Yellow
Material Polypropylene
Type Floor Stand