Learn More
Thermo Scientific™ Flat PCR Caps, strips of 12
Product Code. 12154681
Change view
Click to view available options
Color:
Natural
Product Type:
Flat Cap, 12-Cap Strip
PCR Plate Cap Strip
Unit Size:
Pack of 200
Pack of 80
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Product Type Unit Size
12154681 Natural Flat Cap, 12-Cap Strip Pack of 80
12109840 Natural PCR Plate Cap Strip Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 12154681 Supplier Thermo Scientific™ Supplier No. AB0850
In Stock
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Domed cap strips for PCR applications for use with Thermo Scientific 96-well PCR plates and strip tubes.
Ensure proper sealing on all PCR strip tubes and 96-well PCR plates with precision molded Thermo Scientific™ PCR Plate Cap Strips.
Highlights
  • Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
  • Large end tabs for strip removal (12 strips only)
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
Certifications/Compliance Certified DNA/RNase Free
Material Polypropylene
Quantity 80 strips
Format 80 Strip Caps
For Use With (Equipment) Thermal Cycler
Product Type Flat Cap, 12-Cap Strip