Learn More
Lenz Laborglasinstrumente™ Flat Flange Lid with Center Neck and Two Angled Side Necks
Product Code. 11552354
Change view
Click to view available options
Height (Metric):
75 mm
85 mm
95 mm
110 mm
125 mm
150 mm
165 mm
Includes:
Laboratory Flange 100
Laboratory Flange 140
Laboratory Flange 150
Laboratory Flange 200
Laboratory Flange 60
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Includes Unit Size
11552354 125 mm Laboratory Flange 150 Each
10311560 110 mm Laboratory Flange 100 Each
10775695 95 mm Laboratory Flange 100 Each
11562354 150 mm Laboratory Flange 140 Each
11572354 150 mm Laboratory Flange 200 Each
11582354 165 mm Laboratory Flange 200 Each
11542354 75 mm Laboratory Flange 60 Each
10192454 85 mm Laboratory Flange 60 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11552354 Supplier Lenz Laborglasinstrumente™ Supplier No. 06103444
  • ONLINE EXCLUSIVE PRICE
    423.00 EUR
    323.52 EUR / Pack of 1
    Save 99.48 EUR
    23% Off
Estimated Shipment: 07-10-2026
Specifications
Material Borosilicate Glass, Duran Glass
For Use With (Application) Reaction vessels
Height (Metric) 125 mm
Includes Laboratory Flange 150
Product Type Flange lid