Learn More
Paragon Scientific™ Flash Point Certified Reference Material
Product Code. 15862517
Change view
Click to view available options
Product Type:
COC ASTM D92 ( 153.0 °C), 3 x 80 mL
Unit Size:
Pack of 3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
15862517 COC ASTM D92 ( 153.0 °C), 3 x 80 mL Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15862517 Supplier Paragon Scientific™ Supplier No. CRMUCOCMID
 Restricted Item
Estimated Shipment: 28-12-2026
These are certified reference materials characterized by a method specific multi-laboratory study in accordance with ASTM D92. These materials are suitable for verifying the performance of an instrument and/or procedure as specified in this standard.
  • Certified Reference Material for annual verification checks in accordance with ASTM D92.
  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation by means of a method specific inter-laboratory study.
  • Packaged in 3 x 80 mL bottles – ‘One shot’, one bottle per test ensures zero contamination/ high-end volatile loss.
  • Fully traceable to international standards.
  • Supplied with MSDS.
  • Tamper evident security packaging.
  • 12 month shelf life.
  • Manufactured in the United Kingdom
Specifications
Product Type COC ASTM D92 ( 153.0 °C), 3 x 80 mL
Certifications/Compliance Certified Reference Material for annual verification checks in accordance with ASTM D92
compliance-icons
Product Identifier
  • CRMU-PMCC-MID, CRMU-COC-MID
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contain: white mineral oil
Nominal values quoted