Learn More
Thermo Scientific™ Fixed Needle Syringes for GC Instruments
Product Code. 10025744 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
0.5 μL
5 μL
10 μL
Needle Gauge:
23 G
23 to 26 G
25 G
26 G
26s G
For Use With (Equipment):
AI/AS1310, TriPlus, AS300, AS2000, AS200/AS800, AI/AS 1610 Autosampler
AI/AS1310, TriPlus, AS3000, AI/AS 1610 Autosamplers
AS3000, AS2000, AI/AS1310, AI/AS 1610 Autosamplers
CTC GC Instruments, AI/AS 1610 Autosamplers
TriPlus Autosampler
TriPlus RSH Autosampler
TriPlus, AS2000 Autosampler
TriPlus, AS2000, AI/AS 1610 Autosamplers
TriPlus, AS3000, AS2000, AS200, AS800, AI/AS 1610 Autosampler
Unit Size:
Each
Pack of 6
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Needle Gauge For Use With (Equipment) unitSize
10025744 10 μL 23 G CTC GC Instruments, AI/AS 1610 Autosamplers Pack of 6
10512265 5 μL 26 G AS3000, AS2000, AI/AS1310, AI/AS 1610 Autosamplers Each
10374653 10 μL 25 G TriPlus, AS3000, AS2000, AS200, AS800, AI/AS 1610 Autosampler Each
10197854 10 μL 23 to 26 G TriPlus, AS2000, AI/AS 1610 Autosamplers Each
10384653 10 μL 26 G TriPlus, AS2000 Autosampler Each
10527694 5 μL 26 G TriPlus Autosampler Each
10746173 10 μL 23 G AI/AS1310, TriPlus, AS300, AS2000, AS200/AS800, AI/AS 1610 Autosampler Each
10721724 10 μL 23 G TriPlus, AS2000 Autosampler Each
10364083 5 μL 23 G CTC GC Instruments, AI/AS 1610 Autosamplers Each
10029192 10 μL 26 G CTC GC Instruments, AI/AS 1610 Autosamplers Pack of 6
10206824 10 μL 26 G CTC GC Instruments, AI/AS 1610 Autosamplers Each
10361505 0.5 μL 23 G AI/AS1310, TriPlus, AS3000, AI/AS 1610 Autosamplers Each
13456806 5 μL 26s G TriPlus RSH Autosampler Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 10025744 Supplier Thermo Scientific™ Supplier No. 365D2971
Chromatography - Monthly Promotion
Use promo code 28105
View all promotions
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
Get accurate, reproducible results from these fixed-needle syringes for Thermo Scientific™ and Agilent GC autosamplers.
Get accurate, reproducible fixed-needle syringes for Thermo Scientific™ and Agilent GC autosamplers. Choose from a variety of sizes, types and autosampler compatibilities. They are available in standard industry size with white scale backing for easy reading and solvent-resistant markings.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Needle Gauge 23 G
Length (Metric) 50 mm
Product Type GC Autosampler Syringe, Fixed Needle
Disposable Reusable
Quantity 6/Pk.
Volume (Metric) 10 μL
Length (Metric) Needle 50 mm
Needle Point Style Cone
For Use With (Equipment) CTC GC Instruments, AI/AS 1610 Autosamplers