Learn More
Thermo Scientific™ Removable-Needle Autosampler Syringes for GC Instruments
Shop All Thermo Scientific ProductsDescripción
Thermo Scientific™ Removable-Needle Autosampler Syringes for GC Instruments are accurate and reproducible over a range of volumes.
- Borosilicate glass with metal flanges
- Needles and plungers are stainless steel
- Field-proven syringe design
Especificaciones
Especificaciones
| Material | Stainless Steel (Needle), Stainless Steel (Plunger) |
| Description | 23 to 26 gauge; 5uL; 42mm length; 6/Pk. |
| Needle Gauge | 23 to 26s G |
| Length (Metric) | 42 mm |
| Product Type | GC Autosampler Syringe, Fixed Needle |
| Disposable | Reusable |
| Quantity | 6 Pk. |
| Volume (Metric) | 5 μL |
| Length (Metric) Needle | 42 mm |
| Needle Point Style | Cone |
| Mostrar más |